Order Entry
Switzerland
ContactUsLinkComponent
 
Filament packaging tapes, 8981, Scotch®
  817-0452
 :  3M
undefined
Filament packaging tapes, 8981, Scotch®
  817-0452
 :  3M

 

  • Width:
    50 mm
  • Description:
    Filament tape, with synthetic rubber adhesive
  • Seq:
    02
  • Pk:
    1 Roll

 

 

These high performance 8981 filament packaging tapes are made of Scotchpro™ PP film reinforced with continuous glass yarn and synthetic rubber adhesive.

  • Backing provides excellent resistance to nicks, abrasion and moisture
  • High shear adhesion for low unit cost
  • Heavy-duty closure of hard-to-adhere or high recycled content fiberboard

Length: 50,0 m

Colour: Transparent