Orders LinkContactUsLinkComponent
Laboratory bottles, narrow neck, with thread
 :   Brand
Laboratory bottles, narrow neck, with thread
 :  Brand
Laboratory bottles, narrow neck, with thread
Laboratory bottles, narrow neck, with thread
Soda-lime glass, amber, coated or uncoated.
  • Good chemical and physical properties
  • Maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 °C
DIN thread. Space saving in storage and transport. Stable, non-slip. Amber to protect light-sensitive materials. The coating envelops the glass bottle like a protection skin. Should the bottle break, the splintering hazards are reduced considerably.
Product Family Options
Coated
  • Capacity
    Thread
    Ext.Ø
    Height
    W×D×H
    Colour
  • 100 ml
    GL 32
    125 mm
    50×50×125 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 12.80
    SFr. 12.80
    1 * 1 ST
       SFr. 12.80
  • 250 ml
    GL 32
    160 mm
    65×65×165 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 17.00
    SFr. 17.00
    1 * 1 ST
       SFr. 17.00
  • 500 ml
    GL 32
    195 mm
    80×80×195 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 17.70
    SFr. 17.70
    1 * 1 ST
       SFr. 17.70
  • 1000 ml
    GL 45
    230 mm
    95×95×230 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 21.40
    SFr. 21.40
    1 * 1 ST
       SFr. 21.40
  • 2500 ml
    GL 45
    140 mm
    300 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 39.20
    SFr. 39.20
    1 * 1 ST
       SFr. 39.20
Uncoated
  • Capacity
    Thread
    Ext.Ø
    Height
    W×D×H
    Colour
  • 100 ml
    GL 32
    125 mm
    50×50×125 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 6.80
    SFr. 6.80
    1 * 1 ST
       SFr. 6.80
  • 250 ml
    GL 32
    160 mm
    65×65×160 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 8.30
    SFr. 8.30
    1 * 1 ST
       SFr. 8.30
  • 500 ml
    GL 32
    195 mm
    80×80×195 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 8.70
    SFr. 8.70
    1 * 1 ST
       SFr. 8.70
  • 1000 ml
    GL 45
    230 mm
    95×95×230 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 10.20
    SFr. 10.20
    1 * 1 ST
       SFr. 10.20
  • 2500 ml
    GL 45
    140 mm
    300 mm
    Amber
    1 * 1 ST
    1 * 1 ST - SFr. 22.10
    SFr. 22.10
    1 * 1 ST
       SFr. 22.10