

Tetrachloroethylene ≥99.8%, ULTRA RESI-ANALYZED® for organic trace analysis, for IR spectrophotometry, J.T.Baker®
Catalog # 9360-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.8 %
- Assay (Cl₂C:CCl₂) (by GC):≥99.8 %
- Infrared Absorbance Test: Hydrocarbon (exclusive of stabilizer):≤5 ppm
- Residue after Evaporation:≤10 ppm
- Chloride (Cl):≤1.0 ppm
- Titrable Acid (µeq/g):≤0.5
- Water (H₂O)(by Karl Fischer titrn):≤0.005 %
- Free Halogen (ASTM-D 4755):Passes Test
Specification Test Results
About this item
:
:165.83 g/mol
Merck Index:12,09332
Boiling Pt:121.1 °C (1013 hPa)
Melting Pt:–22 °C
Density:1.63 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-825-9
UN:1897
ADR:6.1,III
:Packaged under inert gas.
:GHS08
:GHS09
CAS Number:127-18-4
Formula:C₂Cl₄
MDL Number:MFCD00000834