ContactUsLinkComponent
Barrier tapes, 766
Barrier tapes, 766
Catalog # 819-0123
Supplier:  3M
undefined
Barrier tapes, 766
Catalog # 819-0123
Supplier:  3M
Specifications
  • Width:
    50,8 mm
  • Description:
    Barrier tape, hazard warning, rubber adhesive
  • Pk:
    24 Roll
Specifications
About this item
These 766 barrier tapes are made of vinyl with a rubber adhesive and are ideal for hazard warning.
  • Conformable backing ideal for use on curved and irregular surfaces
  • Maintains vivid colour over time for long-lasting quality
  • Rubber adhesive provides good instant adhesion on many surfaces
Used for marking of equipment pinch points or gears for warning purposes, temporary floor marking around equipment or spills.
Length: 33,0 m