Condensers, high efficient, four coil
Anbieter: Lenz Laborglas GmbH & CO.KG
Zertifikate
Über diesen Artikel
DURAN® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
High-efficient condensers with four cooling coils, cooling surface: 2,350 cm².
Frequently Bought Together