Test tubes, with spout
Supplier: HIRSCHMANN
Certificates
About this item
DURAN®, borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
- Graduated
Frequently Bought Together