ContactUsLinkComponent
Microfiber wipers for delicate tasks, KIMTECH SCIENCE*
Microfiber wipers for delicate tasks, KIMTECH SCIENCE*
Catalog # 115-0561
Supplier:  KIMBERLY CLARK
Microfiber wipers for delicate tasks, KIMTECH SCIENCE*
Catalog # 115-0561
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    10 wipers per unit
  • Colour:
    White
  • L×W:
    102×76 mm
  • Pk:
    600
Specifications
About this item
These Microfiber Wipers are designed for high end optical lens cleaning in a laboratory environment. They are are heavier than the original KIMTECH SCIENCE* Wipers, so they are ideal for equipment or surface cleaning where more strength is needed. Thanks to their material they will not scratch delicate surfaces.
  • Excellent water and oil absorbency (absorb 4,5 times their weight in oil)
  • Excellent wet and dry strength: Suitable for cleaning surfaces and equipment
  • Extremely low lint
  • No surface streaking or smearing
Colour: White
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904