Orders LinkContactUsLinkComponent
Funnels with short stem
Funnels with short stem
Supplier:   Brand
Funnels with short stem
Supplier:  Brand
Certificates
Funnels with short stem
Funnels with short stem
7 Options Available Below
Product Details & Documents
PP, translucent.
  • External flutings allow air to escape during filtration
  • Excellent chemical resistance, autoclavable
  • Suitable for use with standard filter papers
For rapid filtration.
Product Family Options
Product Information
  • Stem length
    Stem O-Ø
    Funnel O-Ø
    Availability
    Price
  • 44 mm
    5 mm
    45 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 95.10
    SFr. 95.10
    1 * 20 ST
       SFr. 95.10
  • 63 mm
    9 mm
    65 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 133.00
    SFr. 133.00
    1 * 20 ST
       SFr. 133.00
  • 69 mm
    10 mm
    80 mm
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - SFr. 173.00
    SFr. 173.00
    1 * 20 ST
       SFr. 173.00
  • 82 mm
    11 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.90
    SFr. 9.90
    1 * 1 ST
       SFr. 9.90
  • 85 mm
    11 mm
    120 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.30
    SFr. 11.30
    1 * 1 ST
       SFr. 11.30
  • 115 mm
    14 mm
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 18.90
    SFr. 18.90
    1 * 1 ST
       SFr. 18.90
  • 143 mm
    14 mm
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 24.70
    SFr. 24.70
    1 * 1 ST
       SFr. 24.70