Adapters with stopcock and connector
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion
With standard ground socket, standard ground stopcock with solid glass key, bore size 2,5 mm and a straight or right-angled tubing connector. The tubing connector has an external diameter of 8 mm.
Frequently Bought Together