ContactUsLinkComponent
Gas inlet tubes with valve stopcock
  201-2612
undefined
Gas inlet tubes with valve stopcock
  201-2612
  • Cone size:
    14/23 NS
  • Pk:
    1
DURAN®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With valve stopcock for dosing, with glass olive (angle: 90°), with cone NS14/23, tube length from top of the joint: approx. 150 mm.