3-Methylpentane, Supelco®
: Merck
:96-14-0
:
:C₆H₁₄
:MFCD00009342
:86.18 g/mol
:64 °C (1013 hPa)
:–118 °C
:0.664 g/cm³ (20 °C)
:–40 °C
:1208
:3,II
:M-Clarity™ quality level = MQ100
:202-481-4
:GHS02
:GHS08
:GHS07
:GHS09
:Ambient
Frequently Bought Together