



2-Methylbutane, Uvasol® for spectroscopy, Supelco®
1.06056.1000
: Merck
:
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Solubility:0.048 g/L (20 °C)
- Melting Point:-160 °C
- Molar Mass:72.15 g/mol
- Boiling Point:28 °C (1013 hPa)
- Vapor Pressure:800 hPa (20 °C)
- Flash Point:-57 °C
- Explosion Limit:1.3 - 7.6%(V)
- Density:0.62 g/cm³ (20 °C)
- pH:(H₂O) no data available
- Ignition Point:420 °C
- Evaporation Number:1
- Hazard Class:3
- Hazard Class Description:Flammable liquid
:
:72.15 g/mol
:28 °C (1013 hPa)
:–160 °C
:0.62 g/cm³ (20 °C)
:–51 °C
:Refrigerator
:201-142-8
:1265
:3,I
:M-Clarity™ quality level = MQ100
:GHS02
:GHS08
:GHS07
:GHS09
:78-78-4
:CH₃CH₂CH(CH₃)₂
:MFCD00009338