Stopcocks for burette, straight
Catalog # LENZ02700501
Supplier: Lenz Laborglas GmbH & CO.KG
Specifications
- Hole Ø:1,5 mm
- Pk:1
- Spigot:12,5 NS
Specifications
About this item
NS stopcock made of borosilicate glass 3.3, facetted tip with plug of PTFE and screwthread retaining device.
- Excellent chemical resistance
- Excellent temperature resistance