ContactUsLinkComponent
Stopcocks, three-way, Czako
Stopcocks, three-way, Czako
Catalog # LENZ02631804
undefined
Stopcocks, three-way, Czako
Catalog # LENZ02631804
Specifications
  • Hole Ø:
    2,5 mm
  • Description:
    Stopcock, three-way, with PTFE plug
  • Pk:
    1
  • Spigot:
    18,8 NS
Specifications
About this item
Stopcocks, made of DURAN® borosilicate glass.
  • Good temperature resistance
  • Good chemical resistance
With PTFE plug and screwthread retaining device.
Certifications: Leakage rate according to DIN 12540-1.