



2,2,4-Trimethylpentane ≥98%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:540-84-1
:
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MDL Number:MFCD00008943
:114.23 g/mol
Merck Index:13,05212
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
EINECS:208-759-1
UN:1262
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay:Min. 98 %
- Refractive index (20°C):1.391 - 1.393
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 30L Metal drum for liquids UN X (ZPACK12124125) 5L Tin bottle for liquids UN Y (ZPACK12321007)
Packaging Information
spec_VWRC28775.290 spec_VWRC28775.290 spec_VWRC28775.368 spec_VWRC28775.368 spec_VWRC28775.368
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Frequently Bought Together