ContactUsLinkComponent
Hydrophilic interaction chromatography columns (HILIC), Asahipak NH2P
 :  Resonac
Hydrophilic interaction chromatography columns (HILIC), Asahipak NH2P
 :  Resonac
  • Catalog No:
  • 554-2234
  • 554-0878
  • 554-2236
  • 554-0880
  • 554-1776
  • 554-4502
  • 554-1773
  • 554-4437
  • 554-0879
  • Description:
  • Asahipak NH2P-50G 4A
  • Asahipak NH2P-50 2D
  • Asahipak NH2P-50G 2A
  • Asahipak NH2P-50 4E
  • Asahipak NH2P-40 3E
  • Asahipak NH2P-40 2E
  • Asahipak NH2P-50G 3A
  • Asahipak NH2P-50 4B
  • Asahipak NH2P-50 4D
  • Max. pressure:
  • 10 MPa (100 bar)
  • 15 MPa (150 bar)
  • 13 MPa (130 bar)
  • 20 MPa (200 bar)
  • 5,0 MPa (50 bar)
  • 10 MPa (100 bar)
  • pH range:
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • Temperature limit:
  • 4 - 50 °C
  • 4 - 50 °C
  • 4 - 50 °C
  • 4 - 50 °C
  • 15 - 50 °C
  • 15 - 50 °C
  • 15 - 50 °C
  • 4 - 50 °C
  • 4 - 50 °C
  • USP code:
  • L82
  • L82
  • L82
  • L82
  • L82
  • L82
  • L82
  • L82
  • L82
Frequently Bought Together