Sleeves for conical ground joints
Supplier: Lenz Laborglas GmbH & CO.KG
Certificates
About this item
Disposable ground joint sleeves. thin walled, made of PTFE.
- Sleeves have a wall thickness of 0,05 mm
- Usable to 260 °C
- Outstanding chemical resistance
Frequently Bought Together