4-Methyl-2-pentanone ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
CAS Number:108-10-1
:
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:Min. 98 %
- IR Spectrum:Passes test
- Boiling point:115 - 117 °C
- Density (20/4):0.790 - 0.810
- Evaporation residue:Max. 0.02 %
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
spec_VWRC25656.298 spec_VWRC25656.298 spec_VWRC25656.298 spec_VWRC25656.323 spec_VWRC25656.323
Product specification
More
Frequently Bought Together