4-Methyl-2-pentanone, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
Synonyms: Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
CAS Number:108-10-1
:
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938
:100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay (by GC) (corrected for water):min. 99 %
- Appearance:clear
- Color (APHA):max. 15
- Residue after Evaporation:max. 0.002 %
- Titrable Acid (meq/g):max. 0.002
- Water (H2O):max. 0.1 %
- Aluminium (Al):max. 0.5
- Boron (B):max. 0.02
- Barium (Ba):max. 0.1
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.05
- Cobalt (Co):max. 0.02
- Chromium (Cr):max. 0.02
- Copper (Cu):max. 0.02
- Iron (Fe):max. 0.1
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.02
- Nickel (Ni):max. 0.02
- Lead (Pb):max. 0.1
- Tin (Sn):max. 0.1
- Zinc (Zn):max. 0.1
Specification Test Results
Frequently Bought Together