Order Entry
Switzerland
ContactUsLinkComponent
4-Methyl-2-pentanone VLSI for microelectronic, J.T.Baker®
Flammable
Irritant
4-Methyl-2-pentanone VLSI for microelectronic, J.T.Baker®
Catalog # 5384-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
4-Methyl-2-pentanone VLSI for microelectronic, J.T.Baker®
Catalog # 5384-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    8 pt US
  • Seq:
    8026

Specifications

Specification Test Results

  • For Microelectronic Use:
    Passes Test
  • Appearance:
    Passes Test
  • Assay (CH₃COCH₂CH(CH₃)₂) (by GC, corrected for water):
    ≥98.5 %
  • Color (APHA):
    ≤15
  • Density (g/mL) at 25°C:
    0.794 - 0.798
  • Residue after Evaporation:
    ≤0.0050 %
  • Acidity (as CH₃COOH):
    ≤0.01 %
  • Water (H₂O)(by Karl Fischer titrn):
    ≤0.1 %
  • Particle Count - 0.5 µm and greater:
    ≤130 par/ml
  • Particle Count - 1.0 µm and greater:
    ≤10 par/ml
  • Boiling Range (Initial):
    ≥114.0 °C
  • Boiling Range (Dry Point):
    ≤117.0 °C

Specification Test Results

About this item

 :
 :100.16 g/mol
Merck Index:13,05227
Boiling Pt:117.4 °C (1013 hPa)
Melting Pt:–84 °C
Density:0.801 g/cm³ (20 °C)
Flash Pt:16 °C
Storage Temperature:Ambient
EINECS:203-550-1
UN:1245
ADR:3,II
 :GHS02
 :GHS07
CAS Number:108-10-1
Formula:(CH₃)₂CHCH₂COCH₃
MDL Number:MFCD00008938