Order Entry
Switzerland
ContactUsLinkComponent
n-Undecane for synthesis, Sigma-Aldrich®
n-Undecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
n-Undecane for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -26 °C
  • Molar Mass:
    156.31 g/mol
  • Boiling Point:
    195 °C (1013 hPa)
  • Vapor Pressure:
    0.6 hPa (20 °C)
  • Flash Point:
    60 °C
  • Explosion Limit:
    0.6 - 6.5 %(V)
  • Density:
    0.74 g/cm3 (20 °C)
  • Ignition Point:
    220 °C
  • Viscosity Kinematic:
     - 7 mm2/s (40 °C)

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together