

n-Undecane
Catalog # EHERC17896300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
About this item
:
:156.31 g/mol
Boiling Pt:196 °C (1013 hPa)
Melting Pt:–26.7 °C
Density:0.7416 g/cm³ (20 °C)
Flash Pt:60 °C
Storage Temperature:Ambient
EINECS:214-300-6
UN:2330
ADR:3,III
:GHS02
:GHS08
CAS Number:1120-21-4
Formula:H₃C(CH₂)₉CH₃
MDL Number:MFCD00008959