Supelco® Organic Reference Standard, Pentane
Pentane, Analytical standard, 50 mL
Catalog # 76870-50ML
Supplier: Merck
Jump to:
Product Details & Documents
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P273 - P301 + P310 + P331
Specifications
Specifications
- Pk:50 mL
- Standards:Analytical standard
- Description:Pentane
- Boiling point:35 - 36 °C (lit.)
- Hazard class description:H225 - H304 - H336 - H411
- MDL No.:MFCD00009498
- Density:0,626 g/ml at 25 °C (lit.)
- Molecular weight:72,15
- CAS No.:109-66-0
- Cat. No.:76870-50ML
- M-Clarity quality level:MQ100
- Formula:CH₃(CH₂)₃CH₃
- Storage conditions:2...30 °C
- Flash point:–40 °C
- Melting point:–130 °C (lit.)
- Shipping conditions:STD



