Order Entry
Switzerland
ContactUsLinkComponent
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Irritant
Health
Flammable
Environment
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates
undefined
Irritant
Health
Flammable
Environment
n-Pentane ≥99.0%, CHROMASOLV™ for HPLC, Riedel-de Haën™
Certificates

Specification Test Results

  • Assay (GC):
    Min. 99.0 %
  • IR spectrum:
    complying
  • Refractive index (n 20/D):
    1.356 - 1.360
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.005 %
  • Free acid (as CH₃COOH):
    Max. 0.0005 %
  • Fluorescence (chinin) at 254 nm:
    Max. 1.0 ppb
  • Fluorescence (chinin) at 365 nm:
    Max. 1.0 ppb
  • Absorbance at 250 nm:
    Max. 0.004
  • Absorbance at 230 nm:
    Max. 0.02
  • Absorbance at 220 nm:
    Max. 0.07
  • Absorbance at 210 nm:
    Max. 0.20
  • Absorbance at 200 nm:
    Max. 0.70
  • Explosion Limit:
    ~8.3 %
  • Autoignition Temperature:
    500 °F

Specification Test Results

Related Information

Frequently Bought Together