Order Entry
Switzerland
ContactUsLinkComponent
n-Pentane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Health
Irritant
Environment
n-Pentane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9333-03
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Health
Irritant
Environment
n-Pentane ≥99.0% (by GC, corrected for water content), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9333-03
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 5
  • FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤5
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):
    ≤5
  • Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water):
    ≥99.0 %
  • Color (APHA):
    ≤10
  • Residue after Evaporation:
    ≤1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤100 ppm

Specification Test Results

About this item

 :
 :72.15 g/mol
Merck Index:13,07193
Boiling Pt:35…36 °C (1013 hPa)
Melting Pt:–130 °C
Density:0.63 g/cm³ (20 °C)
Flash Pt:–40 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
 :Packaged under inert gas.
REACH:01-2119459286-30
 :GHS02
 :GHS08
 :GHS07
 :GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498