Order Entry
Switzerland
ContactUsLinkComponent
n-Pentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Flammable
Health
Irritant
Environment
n-Pentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Health
Irritant
Environment
n-Pentane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-245 nm:
    ≤0.01
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:
    ≤0.10
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:
    ≤0.4
  • Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:
    ≤190
  • Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:
    ≤0.5 ppb
  • Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:
    ≤1.5 ppb
  • Assay (CH₃(CH₂)₃CH₃) (by GC, corrected for water):
    ≥99.0 %
  • Residue after Evaporation:
    ≤1.0 ppm
  • Substances Darkened by H₂SO₄:
    Passes Test
  • Water (by KF, coulometric):
    ≤0.01 %

Specification Test Results

Related Information

Frequently Bought Together