Order Entry
Switzerland
ContactUsLinkComponent
n-Pentane ≥99% for HPLC
Flammable
Health
Irritant
Environment
n-Pentane ≥99% for HPLC
Certificates
undefined
Flammable
Health
Irritant
Environment
n-Pentane ≥99% for HPLC
Certificates

Specification Test Results

  • Appearance (Color):
    Colorless
  • Form:
    Liquid
  • Color (APHA):
    ≤10
  • Assay (GC):
    ≥98.0% ( as n-Pentane)
  • Comment:
    Sub micron filtered
  • Water Content (Karl Fischer Titration):
    ≤0.02%
  • Optical Absorbance at 200 nm:
    ≤1.0 AU
  • Optical Absorbance at 210 nm:
    ≤0.5 AU
  • Optical Absorbance at 220 nm:
    ≤0.15 AU
  • Optical Absorbance at 245 nm:
    ≤0.01 AU
  • Residue on Evaporation:
    ≤5 ppm

Specification Test Results

Related Information

Frequently Bought Together