One-piece fittings with integrated ferrules
Supplier: S.C.A.T.
Certificates
About this item
These PFA / PTFE fittings connect SCAT SafetyCaps and SafetyWasteCaps to ensure that capillaries and tubes are sealed tightly.
- Made from PFA or PTFE
- Available in different diameters and colours
- Autoclavable
- High flexibility
Suitable for SafetyCaps and SafetyWasteCaps.
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
TDS Universal Waste Hub Jan
Datasheet
Frequently Bought Together