Microbiological filtration system, MBS II, Whatman™
Référence 514-0054
Fournisseur: Whatman products (Cytiva)
Spécifications
- Type:MBS II
- Description:NC membrane, 0,45 µm, black, white grid
- Pk:24
Spécifications
À propos de cet article
The microbiological filtration system is used for optimal microbiological control using membranes. The design of the system, which consists of an electrical membrane dispenser, a funnel dispenser, and a vacuum manifold, leads to more reproducible results.
- Shorter preparation time
- High reproducibility
- Large funnel capacity for foaming liquids
- Funnels can be autoclaved up to 50 times
The special sealing technique ensures easy handling and a good integrity of the funnel and membrane during filtration. This reduces any cross contamination to a minimum.