n-Octane, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:111-65-9
:
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556
:114.23 g/mol
Merck Index:13,06782
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–57 °C
Density:0.703 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-892-1
UN:1262
ADR:3,II
:M-Clarity™ quality level = MQ100
Flash Pt:8 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together