



n-Octane for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:111-65-9
:
Formula:H₃C(CH₂)₆CH₃
MDL Number:MFCD00009556
:114.23 g/mol
Merck Index:13,06782
Boiling Pt:126 °C (1013 hPa)
Melting Pt:–57 °C
Density:0.703 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:203-892-1
UN:1262
ADR:3,II
:M-Clarity™ quality level = MQ200
Flash Pt:8 °C
:GHS02
:GHS08
:GHS07
:GHS09
Specification Test Results
- Solubility:0.0007 g/l (20 °C)
- Melting Point:-57 °C
- Molar Mass:114.23 g/mol
- Boiling Point:125 - 126 °C (1013 hPa)
- Vapor Pressure:14 hPa (20 °C)
- Flash Point:13 °C
- Explosion Limit:0.8 - 6.5 %(V)
- Density:0.70 g/cm3 (20 °C)
- Ignition Point:210 °C
Specification Test Results
Related Information
Frequently Bought Together