Organic reference standards, all-rac-alpha-tocopherols, Ph. Eur.
Supplier: EDQM
Certificates
About this item
Specifications
- CAS No.:7695-91-2
- Formula:C₃₁H₅₂O₃
- Description:All-rac-alpha-tocopheryl acetate for peak identification
Specifications
Frequently Bought Together