Order Entry
Switzerland
ContactUsLinkComponent
n-Hexane, anhydrous (max. 0.005% H₂O) ≥97%, AnalaR NORMAPUR® analytical reagent
Flammable
Health
Toxic
Environment
n-Hexane, anhydrous (max. 0.005% H₂O) ≥97%, AnalaR NORMAPUR® analytical reagent
Catalog # 24603.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Health
Toxic
Environment
n-Hexane, anhydrous (max. 0.005% H₂O) ≥97%, AnalaR NORMAPUR® analytical reagent
Catalog # 24603.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Appearance:
    Clear colourless liquid
  • Acidity:
    Max. 0.001 %
  • Thiophene:
    Passes test
  • Colour value:
    Max. 10 APHA
  • Assay:
    Min. 97 %
  • Residue on evaporation:
    Max. 0.0100 %
  • Formaldehyde-sulphuric colouration:
    Max. 90 APHA
  • Benzene:
    Max. 0.01 %
  • Water:
    Max. 0.005 %
  • Al (Aluminium):
    Max. 0.5 ppm
  • B (Boron):
    Max. 0.02 ppm
  • Ba (Barium):
    Max. 0.1 ppm
  • Ca (Calcium):
    Max. 0.5 ppm
  • Cd (Cadmium):
    Max. 0.01 ppm
  • Co (Cobalt):
    Max. 0.02 ppm
  • Cr (Chromium):
    Max. 0.02 ppm
  • Cu (Copper):
    Max. 0.01 ppm
  • Fe (Iron):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.1 ppm
  • Mn (Manganese):
    Max. 0.02 ppm
  • Ni (Nickel):
    Max. 0.01 ppm
  • Pb (Lead):
    Max. 0.01 ppm
  • Sn (Tin):
    Max. 0.1 ppm
  • Zn (Zinc):
    Max. 0.01 ppm

Specification Test Results

About this item

 :
 :86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94,3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
REACH:01-2119480412-44
 :GHS02
 :GHS08
 :GHS06
 :GHS09
CAS Number:110-54-3
 :92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520