



n-Hexane ≥97.0% (by GC), CHROMASOLV™ for HPLC, Riedel-de Haën™
:110-54-3
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD00009520
:86.18 g/mol
:13,04712
:66…69 °C(1.013 hPa)
:–95 °C
:0.660…0.668 g/cm³ (20 °C)
:7.4 %(V)
:203-777-6
:1208
:3,II
:GHS07
:GHS08
:GHS02
:GHS09
:Ambient
- Assay (GC):Min. 97.00 %
- Non-volatile matter:Max. 0.0005 %
- Water (Karl Fischer):Max. 0.010 %
- Free acid (as CH₃COOH):Max. 0.001 %
- Absorbance at 250 nm:Max. 0.01
- Absorbance at 225 nm:Max. 0.1
- Absorbance at 220 nm:Max. 0.7
- Absorbance at 210 nm:Max. 0.3
- Absorbance (JP)(Val.Ref):complying
- IR spectrum:complying
- Peroxides JP:complying
- Vapor Pressure:~132 mmHg ( 20 °C)
- Refractive Index:n20/D 1.375 (lit.)
- Vapor Density:~3 (vs air)
Frequently Bought Together