



n-Hexane ≥95.0% (by GC, corrected for water content) for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Catalog # 9304-03
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle, amber, narrow mouth
- Pk:4 L
- Seq:5162
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.005
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.008
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 220 nm:≤0.08
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 210 nm:≤0.2
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤192
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):≥99.5 %
- Assay (as n-Hexane) (by GC, corrected for water):≥95.0 %
- Residue after Evaporation:≤2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.01 %
Specification Test Results
About this item
:
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–22 °C
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520