ContactUsLinkComponent
n-Hexane, extra dry 96+%, AcroSeal®
Irritant
Health
Environment
Flammable
n-Hexane, extra dry 96+%, AcroSeal®
Certificates
undefined
Irritant
Health
Environment
Flammable
n-Hexane, extra dry 96+%, AcroSeal®
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Water:
    ≤0.002 %
  • GC:
    ≥96.0 %
  • Residue after evaporation:
    ≤0.0005 %
  • Filt.over 0.2 micron PFTE:
    Filtered