



n-Hexane ∼95% for HPLC
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MDL Number:MFCD00009520
:86.18 g/mol
Merck Index:13,04712
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–95 °C
Density:0.659 g/cm³
Flash Pt:–22 °C
EINECS:203-777-6
UN:1208
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.01 %
- UV:at 200 nm A: ≤0.70
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0002 %
- Acidity:≤0.00015 meq/g
- Filtered to 0.2 micrometer:Filtered
Specification Test Results
Frequently Bought Together