



Hexane (mixture of isomers) 95+% for spectroscopy
:92112-69-1
:H₃C(CH₂)₄CH₃
:MFCD02179311
:86.18 g/mol
:13,04712
:69 °C (1013 hPa)
:–95 °C
:0.659 g/cm³
:–22 °C
:203-777-6
:1208
:3,II
:GHS02
:GHS07
:GHS08
:GHS09
:Ambient
- Appearance:Clear liquid
- Water:≤0.02 %
- GC:≥95.0 % (95% n-hexane approximately)
- UV:1 cm cell vs water
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0005 %
- Acidity:≤0.00015 meq/g
Frequently Bought Together