Order Entry
Switzerland
Orders LinkContactUsLinkComponent
n-Heptane

n-Heptane

Glass bottle, 500 mL
Catalog # WAKO085-00156
Flammable
Health
Irritant
Environment
Marketsource
Supplier:   Wako
n-Heptane
Catalog # WAKO085-00156
Supplier:  Wako
Marketsource
CAS Number:  142-82-5
n-Heptane
n-Heptane

Glass bottle, 500 mL

CAS Number:  142-82-5
Price
Inquire For Price
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  high purity solvents
  • CAS Number:142-82-5
  • Formula:H₃C(CH₂)₅CH₃
  • MDL Number:MFCD00009544
  • Molecular Weight:100.20 g/mol
  • Merck Index:13,04679
  • Boiling Pt:98.4 °C (1013 hPa)
  • Melting Pt:–90.6 °C
  • Density:0.6838 g/cm³ (20 °C)
  • Flash Pt:–4 °C
  • Storage Temperature:Ambient
  • EINECS:205-563-8
  • UN:1206
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.