Order Entry
Switzerland
ContactUsLinkComponent
 
 
 
 
 
n-Heptane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
 
undefined
 
 
 
 
n-Heptane ≥99% (by GC), CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
 

 

  • Assay (GC):
    Min. 99 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Autoignition Temperature:
    433 °F
  • Refractive Index:
    n20/D 1.387 (lit.)
  • Determination of P/N:
    ≤5 ng/L
  • Vapor Pressure:
    83 mmHg ( 37.7 °C)
  • Explosion Limit:
    7 %

 

 

Frequently Bought Together