n-Heptane 99+%, pure
:142-82-5
:
:H₃C(CH₂)₅CH₃
:MFCD00009544
:100.20 g/mol
:13,04679
:98 °C (760 mmHg)
:–91 °C
:0.683 g/cm³
:–4 °C
:205-563-8
:1206
:3,II
:GHS07
:GHS08
:GHS09
:GHS02
:Ambient
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3850 to 1.3890 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:-12° to -16° (20°C, 589 nm) (c=1% ethanol)
Frequently Bought Together