Order Entry
Switzerland
ContactUsLinkComponent
n-Eicosane for synthesis, Sigma-Aldrich®
n-Eicosane for synthesis, Sigma-Aldrich®
Catalog # 8.20547.0050
Supplier:  Merck
CAS Number:  
n-Eicosane for synthesis, Sigma-Aldrich®
Catalog # 8.20547.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    34 - 37 °C
  • Molar Mass:
    282.55 g/mol
  • Boiling Point:
    204 - 206 °C (20 hPa)
  • Vapor Pressure:
    1.3 hPa (20 °C)
  • Flash Point:
    176 °C
  • Density:
    0.79 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :282.55 g/mol
Boiling Pt:342.7 °C (1013 hPa)
Melting Pt:34…37 °C
Density:0.932 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:204-018-1
CAS Number:112-95-8
Formula:H₃C(CH₂)₁₈CH₃
MDL Number:MFCD00009344