Sodium octyl sulphate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:142-31-4
:
Formula:C₈H₁₇NaO₄S
MDL Number:MFCD00007470
:232.28 g/mol
Melting Pt:181…183 °C
Density:1.266 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-535-5
Specification Test Results
- Hill Formula:C₈H₁₇NaO₄S
- Chemical formula:C₈H₁₇NaSO₄
- Molar Mass:232.26 g/mol
- Melting Point:195 °C (decomposition)
- pH value:6 - 8 (10 g/l, H₂O, 20 °C)
- Bulk density:470 kg/m³
- Solubility:(20 °C) soluble
- LD 50 oral:LD50 Rat 3200 mg/kg
- Storage class:10 - 13 Other liquids and solids
- WGK:WGK 2 water endangering
Specification Test Results
Frequently Bought Together