Order Entry
Switzerland
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Sodium dodecyl sulphate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Sodium dodecyl sulphate (SDS) ≥99%, Ultrapure Bioreagent, J.T.Baker®
Supplier:  Avantor
Certificates

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    ≥99%
  • Appearance:
    Passes Test
  • Assay (as C₁₂H₂₅OSO₃Na):
    ≥99 %
  • Absorbance of a 3% w/v Aqueous Solution at 280 nm (1-cm path):
    ≤0.1
  • pH of 1% Solution at 25°C:
    5.0 - 8.0
  • Chloride (Cl):
    ≤0.03 %
  • Nitrogen (N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤1 ppm
  • Heavy Metals (as Pb):
    ≤2 ppm
  • Trace Impurities - Iron (Fe):
    ≤1 ppm
  • DNase Activity (None Detected):
    Passes Test
  • RNase Activity:
    Passes Test

Specification Test Results

Related Information

Frequently Bought Together