Sodium dodecyl sulphate (SDS) 85%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate
CAS Number:151-21-3
:
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175
:288.38 g/mol
Merck Index:14,08636
Melting Pt:206 °C
Density:1,1 g/cm³ (20 °C)
Flash Pt:>150 °C
Storage Temperature:Ambient
EINECS:205-788-1
ADR:4.1,III
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Powder or crystals
- Infrared spectrum:Conforms
- GC:≥80 %
Specification Test Results
Frequently Bought Together