

1,2-Ethandithiol ≥99,0% (durch GC) zur Synthese, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:540-63-6
:
Formula:C₂H₆S₂
MDL Number:MFCD00004892
:94.2 g/mol
Merck Index:13,03760
Boiling Pt:146…149 °C (1013 hPa)
Melting Pt:–41 °C
Density:1,374 g/cm³ (20 °C)
EINECS:208-752-3
UN:1228
ADR:3 (6.1),III
:M-Clarity™ Qualitätsstufe = MQ200
Flash Pt:44 °C
:GHS02
:GHS06
Storage Temperature:Raumtemperatur
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:-41 °C
- Molar Mass:94.20 g/mol
- Boiling Point:146 - 149 °C
- Vapor Pressure:5.3 hPa (23 °C)
- Flash Point:44 °C
- Density:1.12 g/cm3 (20 °C)
- Hazard Class:3
- Hazard Class Description:Flammable liquid
Specification Test Results
Frequently Bought Together