Order Entry
Switzerland
ContactUsLinkComponent
1,3-Propanediol for synthesis, Sigma-Aldrich®
1,3-Propanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
1,3-Propanediol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    100 g/l (20 °C)
  • Melting Point:
    -32 °C
  • Molar Mass:
    76.09 g/mol
  • Boiling Point:
    213 - 215 °C (1013 hPa)
  • Vapor Pressure:
     - 0.1 hPa (20 °C)
  • Refractive Index:
    1.4396 (20 °C, 589 nm)
  • Explosion Limit:
    2.5 %(V)
  • Density:
    1.05 g/cm3 (20 °C)
  • pH:
    4.5 - 7.0 (100 g/l, H2O, 20 °C)
  • Ignition Point:
    405 °C

Specification Test Results

Related Information

Frequently Bought Together