Methanol
Marketsource
Nome del fornitore: Roth Carl
Informazioni su questo articolo
Sinonimi: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- Numero CAS:67-56-1
- Formula:CH₃OH
- Numero MDL:MFCD00004595
- Molecular Weight:32.04 g/mol
- IndexMerck:13,05984
- Punto di ebollizione:64.6 °C (1013 hPa)
- Punto di fusione:–98 °C
- Densità:0.7918 g/cm³ (20 °C)
- Punto di infiammabilità:11 °C (closed cup)
- temperatura di conservazione:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
6 varianti
Product Details & Documents
Sinonimi: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- Numero CAS:67-56-1
- Formula:CH₃OH
- Numero MDL:MFCD00004595
- Molecular Weight:32.04 g/mol
- IndexMerck:13,05984
- Punto di ebollizione:64.6 °C (1013 hPa)
- Punto di fusione:–98 °C
- Densità:0.7918 g/cm³ (20 °C)
- Punto di infiammabilità:11 °C (closed cup)
- temperatura di conservazione:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Specifications for Product Family
Specifiche
Specifiche
- Catalog No:
- ROTH8388.1
- ROTH8388.2
- ROTH8388.5
- ROTH8388.6
- ROTH8388.3
- ROTH8388.4
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
- Chem21 Solvent Selection Guide recommended after discussion
Specifiche
Product Family Options
Informazioni sul prodotto
- Pack typePkDisponibilitàPrice
- Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.1Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.2Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.5Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.6Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.3Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource - Specifiche:
Safer Human and Environmental Health MarketsourceMore
Specifications
Cat. No.ROTH8388.4Environmentally PreferableSafer Human and Environmental HealthEnvironmental certificationChem21 Solvent Selection Guide recommended after discussionSafer Human and Environmental Health Marketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



