Order Entry
Switzerland
ContactUsLinkComponent
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Flammable
Toxic
Health
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Toxic
Health
Methanol ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99,7 %
  • Evaporation residue:
    Max. 5 ppm
  • Water:
    Max. 0,1 %
  • Pesticide analysis (Lindane/ECD):
    Max. 5 ng/l

Specification Test Results

Frequently Bought Together