


Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:67-56-1
:
Formula:CH₃OH
MDL Number:MFCD00004595
:32.04 g/mol
Merck Index:13,05984
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3 (6.1),II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
Specifications
Specification Test Results
- Assay:Min. 98.5 %
- Evaporation residue:Max. 0.015 %
Specification Test Results
Related Information
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951065)
Packaging Information
spec_VWRC20903.290 spec_VWRC20903.368 spec_VWRC20903.415 spec_VWRC20903.461 spec_VWRC20903.550
Product specification
More
VWR Chemicals Big Packs and Bulks VWR Denatured Ethanol
Product Literature
Frequently Bought Together